1,8-Octanedithiol 98%

Supplier: Ambeed
Danger

Synonyms: 1,8-Dimercaptooctane

A581536-5G A581536-25G A581536-1G
77197-834EA 39.14 USD
77197-834 77569-110 77572-856
1,8-Octanedithiol 98%
1,8-Octanedithiol

Precaución: For research use only.

Formula: C₈H₁₈S₂
MW: 178.36 g/mol
MDL Number: MFCD00003574
CAS Number: 1191-62-4
UN: 3334
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR