(±)-1,2-Octanediol 96%
Supplier: Ambeed
|
Warning
|
A581213-100G
A581213-10G
A581213-25G
A581213-500G
A581213-5G
77197-824EA
19
USD
77197-824
77197-826
77197-828
77197-830
77197-832
(±)-1,2-Octanediol 96%
(±)-1,2-Octanediol
Precaución: for Research Use Only
Formula:
C₈H₁₈O₂ |
MDL Number:
MFCD00010738 CAS Number: 1117-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...