4-Bromo-5-chloro-2-fluorobenzenesulfonyl chloride 97%

Supplier: Ambeed
Danger

Synonyms: 4-Bromo-5-chloro-2-fluorobenzene-1-sulphonyl chloride

A581212-1G A581212-250MG A581212-5G
77169-936EA 17 USD
77169-936 77169-938 77893-170
4-Bromo-5-chloro-2-fluorobenzenesulfonyl chloride 97%
4-Bromo-5-chloro-2-fluorobenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂FO₂S
MDL Number: MFCD12026216
CAS Number: 1208075-41-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR