4-Aminopiperidin-2-one 2,2,2-trifluoroacetate 98%
Supplier: Ambeed
|
Warning
|
A580676-100MG
A580676-250MG
A580676-5G
77169-926EA
27.81
USD
77169-926
77169-928
77169-930
4-Aminopiperidin-2-one 2,2,2-trifluoroacetate 98%
4-Aminopiperidin-2-one 2,2,2-trifluoroacetate
Precaución: for Research Use Only
Formula:
C₇H₁₁F₃N₂O₃ MW: 228.17 g/mol |
MDL Number:
MFCD17016140 CAS Number: 1523618-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...