4-N-Boc-aminomethylcyclohexanone 96%

Supplier: Ambeed
Warning

Synonyms: 4-(Aminomethyl)cyclohexanone, N-BOC protected, tert-Butyl ((4-oxocyclohexyl)methyl)carbamate, 4-N-Boc-aminomethyl-cyclohexanone

A580313-100MG A580313-250MG A580313-5G A580313-25G A580313-1G
77169-916EA 25.75 USD
77169-916 77169-918 77169-920 77684-434 77684-433
4-N-Boc-aminomethylcyclohexanone 96%
4-N-Boc-aminomethylcyclohexanone

Precaución: For research use only.

Formula: C₁₂H₂₁NO₃
MW: 227.31 g/mol
MDL Number: MFCD03839885
CAS Number: 809273-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR