1,4-Dibromo-2,5-bis(bromomethyl)benzene 98%

Supplier: Ambeed
Danger

A579459-1G A579459-250MG A579459-5G
77197-818EA 73 USD
77197-818 77197-820 77197-822
1,4-Dibromo-2,5-bis(bromomethyl)benzene 98%
1,4-Dibromo-2,5-bis(bromomethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆Br₄
MW: 421.7500 g/mol
MDL Number: MFCD03411745
CAS Number: 35335-16-1
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR