4-Fluoro-2-methoxybenzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4-Fluoro-2-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane, 4-Fluoro-2-methoxyphenylboronic acid pinacol ester

A579134-1G A579134-250MG A579134-5G
77197-812EA 33.99 USD
77197-812 77197-814 77197-816
4-Fluoro-2-methoxybenzeneboronic acid pinacol ester 98%
4-Fluoro-2-methoxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
MDL Number: MFCD18732613
CAS Number: 624741-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR