2-(4-((Bis((1-(tert-butyl)-1H-1,2,3-triazol-4-yl)methyl)amino)methyl)-1H-1,2,3-triazol-1-yl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: BTTAA

A578749-100MG A578749-1MG A578749-250MG A578749-5MG
77197-804EA 427.45 USD
77197-804 77197-806 77197-808 77197-810
2-(4-((Bis((1-(tert-butyl)-1H-1,2,3-triazol-4-yl)methyl)amino)methyl)-1H-1,2,3-triazol-1-yl)acetic acid 97%
2-(4-((Bis((1-(tert-butyl)-1H-1,2,3-triazol-4-yl)methyl)amino)methyl)-1H-1,2,3-triazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₉H₃₀N₁₀O₂
MW: 430.51 g/mol
MDL Number: MFCD29918287
CAS Number: 1334179-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR