6-Fluoro-3-methyl-1H-indole 97%

Supplier: Ambeed
Warning

A577545-100MG A577545-1G A577545-250MG
77197-788EA 97.85 USD
77197-788 77197-790 77197-792
6-Fluoro-3-methyl-1H-indole 97%
6-Fluoro-3-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈FN
MW: 149.16 g/mol
MDL Number: MFCD03095358
CAS Number: 1011484-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR