1-(5-Carboxypentyl)-2,3,3-trimethyl-3H-indol-1-ium bromide 95%
Supplier: Ambeed
|
Warning
|
A577499-100MG
A577499-1G
A577499-250MG
A577499-5G
77169-898EA
52
USD
77169-898
77169-900
77169-902
77169-904
1-(5-Carboxypentyl)-2,3,3-trimethyl-3H-indol-1-ium bromide 95%
1-(5-Carboxypentyl)-2,3,3-trimethyl-3H-indol-1-ium bromide
Precaución: for Research Use Only
Formula:
C₁₇H₂₄BrNO₂ MW: 354.28 g/mol |
MDL Number:
MFCD02063103 CAS Number: 171429-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...