2,6-Dimethylmorpholine hydrochloride 95%

Supplier: Ambeed
Warning

A576751-100G A576751-25G
77169-894EA 271.92 USD
77169-894 77169-896
2,6-Dimethylmorpholine hydrochloride 95%
2,6-Dimethylmorpholine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 151.63 g/mol
MDL Number: MFCD28099212
CAS Number: 80567-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR