Decane-1,10-diyl bis(2-methylacrylate) 97%

Supplier: Ambeed
Warning

A576628-1G A576628-25G A576628-5G
77197-782EA 9 USD
77197-782 77197-784 77197-786
Decane-1,10-diyl bis(2-methylacrylate) 97%
Decane-1,10-diyl bis(2-methylacrylate)

Precaución: for Research Use Only

Formula: C₁₈H₃₀O₄
MDL Number: MFCD00080452
CAS Number: 6701-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR