4-(tert-Butyl)-N-(4-isopropylphenyl)aniline 97%

Supplier: Ambeed
Warning

A575454-1G A575454-250MG A575454-5G
77169-882EA 86.52 USD
77169-882 77169-884 77169-886
4-(tert-Butyl)-N-(4-isopropylphenyl)aniline 97%
4-(tert-Butyl)-N-(4-isopropylphenyl)aniline

Precaución: for Research Use Only

Formula: C₁₉H₂₅N
MW: 267.42 g/mol
MDL Number: MFCD28130382
CAS Number: 782504-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR