PF-04449913 98+%

Supplier: Ambeed
Danger

Synonyms: Glasdegib, 1-((2R,4R)-2-(1H-Benzo[d]imidazol-2-yl)-1-methylpiperidin-4-yl)-3-(4-cyanophenyl)urea

A575113-10MG A575113-1MG A575113-25MG A575113-5MG
77169-874EA 114 USD
77169-874 77169-876 77169-878 77169-880
PF-04449913 98+%
PF-04449913

Precaución: for Research Use Only

Formula: C₂₁H₂₂N₆O
MW: 374.40 g/mol
MDL Number: MFCD25976839
CAS Number: 1095173-27-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR