N,N-Diallyl-4-methylbenzenesulfonamide 95%

Supplier: Ambeed
Warning

A573498-100MG A573498-1G A573498-250MG
77169-864EA 20.6 USD
77169-864 77169-866 77684-425
N,N-Diallyl-4-methylbenzenesulfonamide 95%
N,N-Diallyl-4-methylbenzenesulfonamide

Precaución: For research use only.

Formula: C₁₃H₁₇NO₂S
MW: 251.345 g/mol
MDL Number: MFCD00226469
CAS Number: 50487-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR