Ethyl-5-methylisoxazole-3-carboxylate 98%

Supplier: Ambeed
Warning

A572678-1G A572678-250MG A572678-25G A572678-5G
77197-768EA 15 USD
77197-768 77197-770 77197-772 77197-774
Ethyl-5-methylisoxazole-3-carboxylate 98%
Ethyl 5-methylisoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₃
MDL Number: MFCD00085080
CAS Number: 3209-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR