2-Methyl-5-nitrophenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-5-nitrobenzeneboronic acid

A572459-1G A572459-250MG A572459-25G A572459-5G
77197-754EA 49.44 USD
77197-754 77197-756 77197-758 77197-760
2-Methyl-5-nitrophenylboronic acid 97%
2-Methyl-5-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₄
MW: 180.96 g/mol
MDL Number: MFCD00757435
CAS Number: 100960-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR