9-(4-(Naphthalen-1-yl)phenyl)-10-(naphthalen-2-yl)anthracene 95%

Supplier: Ambeed
Warning

A572028-100MG A572028-1G A572028-250MG
77169-858EA 39.14 USD
77169-858 77169-860 77169-862
9-(4-(Naphthalen-1-yl)phenyl)-10-(naphthalen-2-yl)anthracene 95%
9-(4-(Naphthalen-1-yl)phenyl)-10-(naphthalen-2-yl)anthracene

Precaución: for Research Use Only

Formula: C₄₀H₂₆
MW: 506.63 g/mol
MDL Number: MFCD11977299
CAS Number: 667940-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR