3,4-Dichloro-1-benzothiophene-2-carbohydrazide 97%

Supplier: Ambeed
Warning

A571011-10MG A571011-1MG A571011-25MG A571011-50MG A571011-5MG
77197-740EA 50 USD
77197-740 77197-742 77197-744 77197-746 77197-748
3,4-Dichloro-1-benzothiophene-2-carbohydrazide 97%
3,4-Dichloro-1-benzothiophene-2-carbohydrazide

Precaución: for Research Use Only

Formula: C₉H₆Cl₂N₂OS
MDL Number: MFCD01993639
CAS Number: 350997-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR