(S)-3-(Cbz-amino)-2-piperidone 97%

Supplier: Ambeed
Warning

A569967-100MG A569967-1G A569967-250MG
77197-732EA 9 USD
77197-732 77197-734 77197-736
(S)-3-(Cbz-amino)-2-piperidone 97%
(S)-3-(Cbz-amino)-2-piperidone

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₃
MDL Number: MFCD08166408
CAS Number: 95582-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR