6-Amino-1-methyl-3,4-dihydroquinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A569249-250MG A569249-5G
77197-716EA 39.14 USD
77197-716 77197-718
6-Amino-1-methyl-3,4-dihydroquinolin-2(1H)-one 97%
6-Amino-1-methyl-3,4-dihydroquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O
MW: 176.21 g/mol
MDL Number: MFCD11987121
CAS Number: 233775-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR