1,3-Bis[(trimethylsilyl)ethynyl]benzene 98+%

Supplier: Ambeed
Warning

Synonyms: 1,3-Bis((trimethylsilyl)ethynyl)benzene

A569187-1G A569187-250MG A569187-5G
77197-710EA 29.87 USD
77197-710 77197-712 77197-714
1,3-Bis[(trimethylsilyl)ethynyl]benzene 98+%
1,3-Bis[(trimethylsilyl)ethynyl]benzene

Precaución: for Research Use Only

Formula: C₁₆H₂₂Si₂
MW: 270.52 g/mol
MDL Number: MFCD00078305
CAS Number: 38170-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR