1-(4-(Methylsulfonyl)phenyl)thiourea 95%

Supplier: Ambeed
Danger

A568802-1G A568802-250MG A568802-5G
77169-852EA 93 USD
77169-852 77169-854 77169-856
1-(4-(Methylsulfonyl)phenyl)thiourea 95%
1-(4-(Methylsulfonyl)phenyl)thiourea

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂S₂
MDL Number: MFCD10693660
CAS Number: 84783-67-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR