2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl 98%

Supplier: Ambeed
Warning

A568646-1G A568646-25G A568646-5G
77197-704EA 9 USD
77197-704 77197-706 77197-708
2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl 98%
2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl

Precaución: for Research Use Only

Formula: C₃₀H₄₁F
MW: 420.656 g/mol
MDL Number: MFCD11053361
CAS Number: 102714-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR