(2S)-(-)-1,1-Bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(S)-DAIPEN, (S)-1,1-Bis(4-methoxyphenyl)-3-methylbutane-1,2-diamine
A566467-1G
A566467-250MG
A566467-100MG
77197-700EA
268.83
USD
77197-700
77197-702
77747-166
(2S)-(-)-1,1-Bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine 98%
(2S)-(-)-1,1-Bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine
Precaución: for Research Use Only
Formula:
C₁₉H₂₆N₂O₂ MW: 314.42 g/mol |
MDL Number:
MFCD02684540 CAS Number: 148369-91-9 UN: 3259 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...