3-Methoxypyrocatechol 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydroxyanisole, 3-Methoxycatechol, 3-Methoxybenzene-1,2-diol

A566396-100G A566396-5G
77197-696EA 281.19 USD
77197-696 77197-698
3-Methoxypyrocatechol 98%
3-Methoxypyrocatechol

Precaución: for Research Use Only

Formula: C₇H₈O₃
MW: 140.14 g/mol
MDL Number: MFCD00002191
CAS Number: 934-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR