3-Bromo-6-fluoro-2-nitrotoluene 96%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-3-fluoro-2-methylnitrobenzene, 1-Bromo-4-fluoro-3-methyl-2-nitrobenzene

A565894-1G A565894-250MG
77169-848EA 40 USD
77169-848 77169-850
3-Bromo-6-fluoro-2-nitrotoluene 96%
3-Bromo-6-fluoro-2-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD18632532
CAS Number: 1286734-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR