1,5-Dichloro-2,4-dinitrobenzene 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
1,3,-Dichloro-4,6-dinitrobenzene
A565581-1G
A565581-25G
A565581-5G
77197-684EA
5.15
USD
77197-684
77197-686
77197-688
1,5-Dichloro-2,4-dinitrobenzene 98%
1,5-Dichloro-2,4-dinitrobenzene
Precaución: for Research Use Only
Formula:
C₆H₂Cl₂N₂O₄ MW: 237 g/mol |
MDL Number:
MFCD00024186 CAS Number: 3698-83-7 UN: 1325 ADR: 4.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...