(S)-(+)-1-Boc-3-fluoropyrrolidine 97%

Supplier: Ambeed
Warning

A565064-1G A565064-250MG
77197-676EA 77 USD
77197-676 77197-678
(S)-(+)-1-Boc-3-fluoropyrrolidine 97%
(S)-(+)-1-Boc-3-fluoropyrrolidine

Precaución: for Research Use Only

Formula: C₉H₁₆FNO₂
MDL Number: MFCD04114064
CAS Number: 479253-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR