1,3-diisopropylurea 95%

Supplier: Ambeed
Warning

A564262-100MG A564262-250MG A564262-5G A564262-1G
77197-666EA 5 USD
77197-666 77197-668 77572-794 77594-718
1,3-diisopropylurea 95%
1,3-diisopropylurea

Precaución: for Research Use Only

Formula: C₇H₁₆N₂O
MDL Number: MFCD00276304
CAS Number: 4128-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR