4-Chloro-6-(chloromethyl)-2-methoxypyrimidine 95%

Supplier: Ambeed
Warning

A564239-100MG A564239-250MG
77169-842EA 61.8 USD
77169-842 77169-844
4-Chloro-6-(chloromethyl)-2-methoxypyrimidine 95%
4-Chloro-6-(chloromethyl)-2-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂O
MW: 193.03 g/mol
MDL Number: MFCD28400977
CAS Number: 94170-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR