(tert-Butoxycarbonylmethyl)triphenylphosphonium chloride 98+%

Supplier: Ambeed
Warning

Synonyms: (2-(tert-Butoxy)-2-oxoethyl)triphenylphosphonium chloride

A562617-100G A562617-1G A562617-25G A562617-5G
77197-652EA 203.94 USD
77197-652 77197-654 77197-656 77197-658
(tert-Butoxycarbonylmethyl)triphenylphosphonium chloride 98+%
(tert-Butoxycarbonylmethyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₄H₂₆ClO₂P
MW: 412.90 g/mol
MDL Number: MFCD00043160
CAS Number: 35000-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR