C-Quinolin-3-ylmethylamine 95%

Supplier: Ambeed
Warning

Synonyms: Quinolin-3-ylmethanamine

A562439-100MG A562439-250MG
77169-830EA 36.05 USD
77169-830 77169-832
C-Quinolin-3-ylmethylamine 95%
C-Quinolin-3-ylmethylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
MDL Number: MFCD00465220
CAS Number: 7521-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR