5-(tert-Butyl)-2-chloropyridine 97%

Supplier: Ambeed
Warning

A561904-1G A561904-250MG A561904-25G A561904-5G
77169-820EA 20 USD
77169-820 77169-822 77169-824 77169-826
5-(tert-Butyl)-2-chloropyridine 97%
5-(tert-Butyl)-2-chloropyridine

Precaución: for Research Use Only

Formula: C₉H₁₂ClN
MDL Number: MFCD28040569
CAS Number: 102236-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR