1,2,3-Tribromobenzene 98%

Supplier: Ambeed
Warning

A561690-100MG A561690-1G A561690-250MG A561690-5G
77197-642EA 9 USD
77197-642 77197-644 77197-646 77551-686
1,2,3-Tribromobenzene 98%
1,2,3-Tribromobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₃
MDL Number: MFCD00192668
CAS Number: 608-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR