(3-Chlorophenyl)methanesulfonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 3-Chlorobenzylsulphonyl chloride, 3-Chloro-α-toluenesulphonyl chloride

A561586-1G A561586-250MG
77169-816EA 56.65 USD
77169-816 77169-818
(3-Chlorophenyl)methanesulfonyl chloride 95%
(3-Chlorophenyl)methanesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
MDL Number: MFCD02683111
CAS Number: 24974-73-0
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR