YH239-EE 98+%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-(2-(tert-butylamino)-1-(N-(4-chlorobenzyl)formamido)-2-oxoethyl)-6-chloro-1H-indole-2-carboxylate, 6-Chloro-3-[1-[[(4-chlorophenyl)methyl]formylamino]-2-[(1,1-dimethylethyl)amino]-2-oxoethyl]-1H-indole-2-carboxylic acid Ethyl Ester

A561517-10MG A561517-1MG A561517-50MG A561517-5MG
77197-628EA 94.76 USD
77197-628 77197-630 77197-632 77197-634
YH239-EE 98+%
YH239-EE

Precaución: for Research Use Only

Formula: C₂₅H₂₇Cl₂N₃O₄
MW: 504.41 g/mol
MDL Number: MFCD28099816
CAS Number: 1364488-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR