6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione 98%

Supplier: Ambeed
Warning

A561230-1G A561230-5G A561230-25G
77197-622EA 17.01 USD
77197-622 77197-624 77197-626 77551-684
6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione 98%
6,6-Dimethyl-5,7-dioxaspiro[2.5]octane-4,8-dione

Precaución: for Research Use Only

Formula: C₈H₁₀O₄
MDL Number: MFCD00042796
CAS Number: 5617-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR