7-Chloro-4-hydroxy-3-(3-phenoxyphenyl)quinolin-2(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: L-701324

A560344-10MG A560344-1MG A560344-25MG A560344-50MG
77197-612EA 100.94 USD
77197-612 77197-614 77197-616 77197-618
7-Chloro-4-hydroxy-3-(3-phenoxyphenyl)quinolin-2(1H)-one 98%
7-Chloro-4-hydroxy-3-(3-phenoxyphenyl)quinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₂₁H₁₄ClNO₃
MW: 363.79 g/mol
MDL Number: MFCD00910917
CAS Number: 142326-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR