Ethyl 1-(pyrimidin-4-yl)-1H-pyrazole-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A560132-100MG
A560132-1G
A560132-250MG
77197-600EA
37.08
USD
77197-600
77197-602
77197-604
Ethyl 1-(pyrimidin-4-yl)-1H-pyrazole-4-carboxylate 97%
Ethyl 1-(pyrimidin-4-yl)-1H-pyrazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₀N₄O₂ MW: 218.21 g/mol |
MDL Number:
MFCD21602179 CAS Number: 1014632-04-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...