1-(1-Benzofuran-2-yl)-2-bromoethan-1-one 96%

Supplier: Ambeed
Danger

Synonyms: 2-(Bromoacetyl)benzo[b]furan, 1-(Benzofuran-2-yl)-2-bromoethanone

A560025-1G A560025-5G
77197-596EA 50.47 USD
77197-596 77197-598
1-(1-Benzofuran-2-yl)-2-bromoethan-1-one 96%
1-(1-Benzofuran-2-yl)-2-bromoethan-1-one

Precaución: for Research Use Only

Formula: C₁₀H₇BrO₂
MW: 239.08 g/mol
MDL Number: MFCD00176644
CAS Number: 23489-36-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR