1-Allylcyclopropane-1-sulfonyl chloride 97%
Supplier: Ambeed
|
Danger
|
A559871-1G
A559871-5G
77197-592EA
15
USD
77197-592
77197-594
1-Allylcyclopropane-1-sulfonyl chloride 97%
1-Allylcyclopropane-1-sulfonyl chloride
Precaución: for Research Use Only
Formula:
C₆H₉ClO₂S |
MDL Number:
MFCD11046295 CAS Number: 923032-59-1 UN: 3265 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...