(1-Methylnaphthalen-4-yl)methanol 96%

Supplier: Ambeed
Warning

Synonyms: (4-Methylnaphthalen-1-yl)methanol

A559211-1G A559211-250MG A559211-25G A559211-5G
77169-804EA 33.99 USD
77169-804 77169-806 77169-808 77684-409
(1-Methylnaphthalen-4-yl)methanol 96%
(1-Methylnaphthalen-4-yl)methanol

Precaución: For research use only.

Formula: C₁₂H₁₂O
MW: 172.223 g/mol
MDL Number: MFCD00155252
CAS Number: 57322-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR