1-(3-(4-Aminophenyl)propanoyl)azetidin-2-one 97%

Supplier: Ambeed
Warning

A558820-100MG A558820-1G A558820-250MG
77197-576EA 39.14 USD
77197-576 77197-578 77242-034
1-(3-(4-Aminophenyl)propanoyl)azetidin-2-one 97%
1-(3-(4-Aminophenyl)propanoyl)azetidin-2-one

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂
MW: 218.252 g/mol
MDL Number: MFCD24627737
CAS Number: 1024869-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR