3-((6-Chloropyridin-2-yl)amino)propanoic acid 97%

Supplier: Ambeed
Warning

A558316-1G A558316-250MG A558316-5G
77197-560EA 210.12 USD
77197-560 77197-562 77197-564
3-((6-Chloropyridin-2-yl)amino)propanoic acid 97%
3-((6-Chloropyridin-2-yl)amino)propanoic acid

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MW: 200.62 g/mol
MDL Number: MFCD18915444
CAS Number: 76629-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR