2-(tert-Butyl)-5-chloro-1H-indole 98%

Supplier: Ambeed
Warning

A556980-100MG A556980-1G A556980-250MG
77169-792EA 109.18 USD
77169-792 77169-794 77169-796
2-(tert-Butyl)-5-chloro-1H-indole 98%
2-(tert-Butyl)-5-chloro-1H-indole

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClN
MW: 207.69 g/mol
MDL Number: MFCD05224151
CAS Number: 69622-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR