(6Z,9Z)-18-Bromooctadeca-6,9-diene 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A556549-100MG A556549-1G A556549-250MG A556549-5G
77169-786EA 54.59 USD
77169-786 77169-788 77169-790 77684-403
(6Z,9Z)-18-Bromooctadeca-6,9-diene 97%, Ambeed.Inc
(6Z,9Z)-18-Bromooctadeca-6,9-diene

Precaución: For research use only.

Formula: C₁₈H₃₃Br
MW: 329.35 g/mol
MDL Number: MFCD11840188
CAS Number: 13044-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR