(S)-(-)-4-Chloro-α-methylbenzyl alcohol 97%

Supplier: Ambeed
Warning

A555841-1G A555841-250MG A555841-5G
77197-546EA 28 USD
77197-546 77197-548 77617-360
(S)-(-)-4-Chloro-α-methylbenzyl alcohol 97%
(S)-(-)-4-Chloro-α-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉ClO
MDL Number: MFCD06797296
CAS Number: 99528-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR