(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)pentanoic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-D-N-Me-Nva-OH
A554246-100MG
A554246-1G
A554246-250MG
77169-768EA
10.3
USD
77169-768
77169-770
77169-772
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)pentanoic acid 95%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)pentanoic acid
Precaución: for Research Use Only
Formula:
C₂₁H₂₃NO₄ MW: 353.41 g/mol |
MDL Number:
MFCD07366866 CAS Number: 1799443-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...