6-Bromo-4-chloronicotinic acid 97%

Supplier: Ambeed
Warning

A553734-100MG A553734-1G A553734-250MG A553734-5G
77169-760EA 37 USD
77169-760 77169-762 77169-764 77169-766
6-Bromo-4-chloronicotinic acid 97%
6-Bromo-4-chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MDL Number: MFCD13189219
CAS Number: 1060808-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR